missing translation for 'onlineSavingsMsg'
Learn More
Pyrazole, 98%, pure, Thermo Scientific Chemicals Product Code.: 10225280

Pyrazole, 98%, pure, Thermo Scientific Chemicals

Product Code. 10225280
25 g, Glass bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10225280

Brand: Thermo Scientific Acros 131740250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-13-1
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005234
InChI Key WTKZEGDFNFYCGP-UHFFFAOYSA-N
Synonym pyrazole, 1,2-diazole, 1h-pyrazol, pyrazol, unii-3qd5kjz7zj, 3qd5kjz7zj, diazole, hpz, 1-h-pyrazole, pyrazol#1
PubChem CID 1048
ChEBI CHEBI:17241
IUPAC Name 1H-pyrazole
SMILES C1=CNN=C1

Specifications

Melting Point 66.0°C to 70.0°C
Color White to Yellow
Boiling Point 186.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 23, 39
Fieser 01,958; 12,419
Merck Index 15, 8071
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 68.08
Percent Purity 98%
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Pyrazole, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0368192 Certificate TypeCertificate of Analysis Date10/01/2023 Catalog Number131740250,131741000
compliance-icons
Product Identifier
  • Pyrazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrazole, 98%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.