missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Amber Borosilicate Glass Class A Certified Volumetric Flask

Manufactured from borosilicate glass with amber coating on exterior surfaces to facilitate use with light sensitive solutions

Brand:  Pyrex 3152/10AWC

231.80 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11393654

  • 251.00 € / Pack of 5
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Calibrated to tolerances required by United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
  • Engraved with individual serial number
  • Supplied with certificate of calibration traceable to National standards
  • Conformity ‘H’ certified
  • Polyethylene stopper
  • White enamel inscriptions
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 1042, DIN 12664, USP, ISO/DIN, Conformity 'H' certified
Amber
50 mL
140 mm
Yes
12/2
Polyethylene
White enamel inscriptions
Borosilicate Glass
±0.05 mL
A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers