Learn More
Pyrex™ Aspirator Bottles, Large
Ideal for storage of distilled water
219.00 € - 494.00 €
Specifications
Includes Lid, Plastic connector and Rubber/PTFE sealing ring
Material Borosilicate Glass
Mouth Wide
Product Type Aspirator Bottle
View More Specs
Products 3
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Neck Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Neck Price Quantity & Availability  
10195034
View Documents
Pyrex
1565/16M
5000 mL 100 mm N/A N/A N/A
12433451
View Documents
Pyrex
1565/22M
20 L 160 mm N/A N/A N/A
11975809
View Documents
Pyrex
1565/18M
10,000 mL 127 mm N/A N/A N/A
Description
Description
  • Wide neck with simple glass cover
  • Supplied with plastic connector and rubber/PTFE sealing ring
  • Accepts either stopcock GP6/10W or glass tubing of 14mm O.D.
Specifications
Specifications
Lid, Plastic connector and Rubber/PTFE sealing ring
Borosilicate Glass
Wide
Aspirator Bottle
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Pyrex™ Aspirator Bottles, Large