Learn More
Pyrex™ Borosilicate Glass Class A Volumetric Flask
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Class A Volumetric Flask includes white enamel inscriptions for easy volume assessment and a polyethylene stopper. Complies with ISO 1042 and DIN 12664, is calibrated to the tolerance required by the United States Pharmacopoeia (USP,) and Conformity 'H' certified.
58.20 € - 223.00 €
Specifications
Certifications/Compliance | ISO 1042, DIN 12664, USP, Conformity 'H' certified |
---|---|
Closure Material | Polyethylene |
Description | White enamel inscriptions |
Material | Borosilicate Glass |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Closure Size | Height (Metric) | Tolerance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Height (Metric) | Tolerance | Price | Quantity & Availability | |||
11303634
|
Pyrex
3151/22 |
1000 mL | 24/29 | 300 mm | ±0.3 mL |
130.00 €
Pack of 2 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||
10517110
|
Pyrex
3151/24 |
2000 mL | 29/32 | 370 mm | ±0.5 mL |
112.00 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||
11363624
|
Pyrex
3151/14 |
100 mL | 14/23 | 170 mm | ±0.08 mL |
152.44 €
Pack of 5 |
In Stock
Log in to see stock. |
|||
11313634
|
Pyrex
3151/01 |
1 mL | 7/16 | 65 mm | ±0.02 mL |
130.00 €
Pack of 5 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||
11323624
|
Pyrex
3151/06 |
10 mL | 10/19 | 90 mm | ±0.02 mL |
102.00 €
Pack of 5 |
In Stock
Log in to see stock. |
|||
11373624
|
Pyrex
3151/16 |
200 mL | 14/23 | 210 mm | ±0.1 mL |
197.00 €
Pack of 5 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||
11333624
|
Pyrex
3151/08 |
20 mL | 10/19 | 110 mm | ±0.03 mL |
78.50 €
Pack of 5 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||
11383624
|
Pyrex
3151/18 |
250 mL | 14/23 | 220 mm | ±0.12 mL |
58.20 €
Pack of 2 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||
11343624
|
Pyrex
3151/10 |
25 mL | 10/19 | 110 mm | ±0.03 mL |
117.00 €
Pack of 5 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||
11393624
|
Pyrex
3151/20 |
500 mL | 19/26 | 260 mm | ±0.2 mL |
99.10 €
Pack of 2 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||
11323634
|
Pyrex
3151/02 |
2 mL | 7/16 | 65 mm | ±0.02 mL |
187.20 €
Pack of 5 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||
10398459
|
Pyrex
3151/26 |
5 L | 34/35 | 475 mm | ±1 mL |
223.00 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||
11353624
|
Pyrex
3151/12 |
50 mL | 12/21 | 140 mm | ±0.05 mL |
118.00 €
Pack of 5 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||
11333634
|
Pyrex
3151/04 |
5 mL | 10/19 | 70 mm | ±0.02 mL |
98.59 €
Pack of 5 |
In Stock
Log in to see stock. ![]() |
|||
Description
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Class A Volumetric Flask includes white enamel inscriptions for easy volume assessment and a polyethylene stopper. Complies with ISO 1042 and DIN 12664, is calibrated to the tolerance required by the United States Pharmacopoeia (USP,) and Conformity 'H' certified.
- Complies with ISO 1042 and DIN 12664
- Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which is tighter than those of ISO/DIN standards
- Each flask is laser etched with an individual serial number batch code and date of manufacture
- Batch certificate available to download online
- Conformity 'H' certified
- Manufactured from chemically resistant Pyrex™ Borosilicate Glass
- Inscriptions in white enamel
- Polyethylene stopper
Specifications
ISO 1042, DIN 12664, USP, Conformity 'H' certified | |
White enamel inscriptions | |
Yes |
Polyethylene | |
Borosilicate Glass | |
A |