Learn More
Pyrex™ Borosilicate Glass Narrow Neck Round Bottom Boiling Flask
Resistant to chemical corrosion and thermal shock
Brand: Pyrex 1100/38D
This item is not returnable.
View return policy
Description
- Complies with ISO 1773
- Manufactured from Pyrex™ borosilicate glass which offers excellent resistance to chemical corrosion and thermal shock
Specifications
ISO 1773 | |
Borosilicate Glass | |
Round | |
62 |
20 L | |
515 mm | |
345 mm |