Learn More
Pyrex™ Borosilicate Glass Narrow Neck Round Bottom Boiling Flask
Resistant to chemical corrosion and thermal shock
Brand:  Pyrex 1100/18D
 View more versions of this product
Product Code. 12009012
  • 171.00 € / Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complies with ISO 1773
  • Manufactured from Pyrex™ borosilicate glass which offers excellent resistance to chemical corrosion and thermal shock
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 1773
Borosilicate Glass
Round
29
500 mL
175 mm
105 mm