Learn More
Pyrex™ Borosilicate Glass Spare Glass Keys
Manufactured from borosilicate glass
Brand:  Pyrex SST103K
 View more versions of this product
Product Code. 15633076
  • 116.00 € / Pack of 5
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Replacement keys for SSD: series stopcocks
  • Manufactured from borosilicate glass
  • Supplied complete with key retaining device
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spare Glass Key
2.5 mm
14.5 mm
Borosilicate Glass