missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Spare Glass Keys
Manufactured from borosilicate glass
32.15 € - 116.00 €
Product Code | Brand | Diameter (Metric) | Plug Bore | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Plug Bore | Price | Quantity & Availability | |||||
15633076
|
Pyrex
SST103K |
14.5 mm | 2.5 mm |
116.00 €
Pack of 5 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
12454761
|
Pyrex
SSD102K |
12.5 mm | 1.5 mm |
32.15 €
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||||
Description
- Replacement keys for SSD: series stopcocks
- Manufactured from borosilicate glass
- Supplied complete with key retaining device
Specifications
Spare Glass Key | |
Borosilicate Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyrex™ Borosilicate Glass Spare Glass Keys