Learn More
Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key
For use when liquids have to be added to systems under vacuum.
Brand:  Pyrex 443554
 View more versions of this product
Product Code. 12915131
  • 183.00 € / Each
Estimated Shipment: 09-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complies with ISO 4800, Type 4
  • With interchangeable PTFE key for grease free performance
  • Double graduated scale showing volume of liquid contained/delivered
  • Stem inside cone for easy fitting to Quickfit™ assemblies
  • Graduations in white enamel
  • Supplied with hollow glass stopper
TRUSTED_SUSTAINABILITY
Specifications
Specifications
500 mL
Borosilicate Glass
Yes
Pressure-equalizing Funnel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key > Capacity: 500mL