Learn More
Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key
For use when liquids have to be added to systems under vacuum.
153.00 € - 183.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
12905131
|
Pyrex
443547 |
250 mL |
153.00 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
12915131
|
Pyrex
443554 |
500 mL |
183.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
12995121
|
Pyrex
443533 |
50 mL |
166.13 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
13431869
|
Pyrex
443540 |
100 mL |
160.00 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
Description
- Complies with ISO 4800, Type 4
- With interchangeable PTFE key for grease free performance
- Double graduated scale showing volume of liquid contained/delivered
- Stem inside cone for easy fitting to Quickfit™ assemblies
- Graduations in white enamel
- Supplied with hollow glass stopper
Specifications
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key