missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Petri Dishes
Manufactured from Pyrex™ borosilicate glass for superior chemical resistance and clarity. Pyrex™ Borosilicate Glass Petri Dishes are a reliable choice for growing bacteria or fungi under laboratory conditions.
Brand: Pyrex 1480/02D
182.23 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Specially formed to ensure even wall thickness and consistent optical performance
- Autoclavable
- Tight fitting lid safeguards clean environment

Specifications
Closure Included | |
Borosilicate Glass | |
Petri Dish | |
55 mm |
Borosilicate Glass | |
Round | |
20 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyrex™ Borosilicate Glass Petri Dishes > Dimensions: 60 dia. x 20mmH
Spot an opportunity for improvement?Share a Content Correction