missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Petri Dishes
Manufactured from Pyrex™ borosilicate glass for superior chemical resistance and clarity. Pyrex™ Borosilicate Glass Petri Dishes are a reliable choice for growing bacteria or fungi under laboratory conditions.
Brand: Pyrex 1480/06D
This item is not returnable.
View return policy
Description
- Specially formed to ensure even wall thickness and consistent optical performance
- Autoclavable
- Tight fitting lid safeguards clean environment
Specifications
Closure Included | |
Borosilicate Glass | |
Petri Dish | |
75 mm |
Borosilicate Glass | |
Round | |
20 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction