Learn More
Pyrex™ Media-lab Bottles with Thermoplastic Polyester Cap
Manufactured from temperature and chemically resistant borosilicate glass. Pyrex™ Media-lab Bottles with Thermoplastic Polyester Cap includes chemically resistant PTFE sealing disc and pouring ring. White graduations and marking spot add easy volume assessment. Withstands hot air sterilization up to 180° C. Complies with ISO 4796.
43.40 € - 564.00 €
Specifications
Includes High temperature cap and pouring ring
Material Borosilicate Glass
Certifications/Compliance ISO 4796
Closure Included Yes
Closure Material Thermoplastic Polyester
View More Specs
Products 8
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
15201499
View Documents
Pyrex
1516/01RED
25 mL 25 mm
129.00 €
Pack of 10
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
15211499
View Documents
Pyrex
1516/02RED
50 mL 32 mm
183.00 €
Pack of 10
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
15221499
View Documents
Pyrex
1516/04RED
100 mL 45 mm
213.00 €
Pack of 10
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
15231499
View Documents
Pyrex
1516/06RED
250 mL 45 mm
221.00 €
Pack of 10
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
15241499
View Documents
Pyrex
1516/08RED
500 mL 45 mm
245.00 €
Pack of 10
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
15251499
View Documents
Pyrex
1516/10RED
1000 mL 45 mm
298.00 €
Pack of 10
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
10660191
View Documents
Pyrex
1516/12RED
2000 mL 45 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15901381
View Documents
Pyrex
1516/12RED
2000 mL 45 mm
564.00 €
Pack of 10
Estimated Shipment: 25-04-2025
to see stock.
Add to basket
Description
Description
  • Manufactured from temperature and chemically resistant borosilicate glass
  • Bottles complete with thermoplastic polyester cap, PTFE sealing disc, and pouring ring
  • Withstands hot air sterilization up to 180° C
  • PTFE sealing disc resists chemical corrosion
  • White graduations and marking spot add easy volume assessment
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
Specifications
Specifications
High temperature cap and pouring ring
ISO 4796
Thermoplastic Polyester
Borosilicate Glass
Yes
Cap
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Pyrex™ Media-lab Bottles with Thermoplastic Polyester Cap