missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key
Supplied with hollow glass stopper and interchangeable PTFE key for grease-free performance. Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key provides good control of the liquid/liquid interface.
Brand: Pyrex 443054N
150.10 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Gilson style with long delivery stem for good control of the liquid/liquid interface
- With interchangeable PTFE key for grease-free performance
- Supplied with hollow glass stopper
Specifications
500 mL | |
Hollow Glass stopper, PTFE key | |
(Bore Size) 4mm |
Borosilicate Glass | |
24/29 | |
Separatory Funnel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction