missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-carboximidamide hydrochloride, 95%, Thermo Scientific™

61.20 € - 449.00 €

Chemical Identifiers

CAS 6345-27-3
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.60
MDL Number MFCD00040385
InChI Key IONKMFGAXKCLMI-UHFFFAOYSA-N
Synonym isonicotinimidamide hydrochloride, pyridine-4-carboximidamide hydrochloride, 4-amidinopyridine hydrochloride, isonicotinamidine hydrochloride, pyridine-4-carboxamidine hydrochloride, 4-pyridinecarboximidamide hydrochloride, 4-pyridinecarboximidamide, monohydrochloride, isonicotinamidine, hydrochloride, 4-amidino-pyridine hydrochloride, pyridine-4-carboxamidine, chloride
PubChem CID 2776844
IUPAC Name pyridine-4-carboximidamide;hydrochloride
SMILES Cl.NC(=N)C1=CC=NC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10712967
View Documents Promotion Details
Thermo Scientific Maybridge
CD05159DE
5 g
61.20 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10661483
View Documents Promotion Details
Thermo Scientific Maybridge
CD05159EA
10 g
101.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10748455
View Documents Promotion Details
Thermo Scientific Maybridge
CD05159EE
50 g
449.00 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

6345-27-3
157.60
IONKMFGAXKCLMI-UHFFFAOYSA-N
2776844
Cl.NC(=N)C1=CC=NC=C1
C6H8ClN3
MFCD00040385
isonicotinimidamide hydrochloride, pyridine-4-carboximidamide hydrochloride, 4-amidinopyridine hydrochloride, isonicotinamidine hydrochloride, pyridine-4-carboxamidine hydrochloride, 4-pyridinecarboximidamide hydrochloride, 4-pyridinecarboximidamide, monohydrochloride, isonicotinamidine, hydrochloride, 4-amidino-pyridine hydrochloride, pyridine-4-carboxamidine, chloride
pyridine-4-carboximidamide;hydrochloride

Specifications

6345-27-3
100.0
C6H8ClN3
isonicotinimidamide hydrochloride, pyridine-4-carboximidamide hydrochloride, 4-amidinopyridine hydrochloride, isonicotinamidine hydrochloride, pyridine-4-carboxamidine hydrochloride, 4-pyridinecarboximidamide hydrochloride, 4-pyridinecarboximidamide, monohydrochloride, isonicotinamidine, hydrochloride, 4-amidino-pyridine hydrochloride, pyridine-4-carboxamidine, chloride
Cl.NC(=N)C1=CC=NC=C1
157.60
157.6
Pyridine-4-carboximidamide hydrochloride
96.5
95%
MFCD00040385
IONKMFGAXKCLMI-UHFFFAOYSA-N
pyridine-4-carboximidamide;hydrochloride
2776844
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications