missing translation for 'onlineSavingsMsg'
Learn More
Pyridine hydrobromide, 98%, Thermo Scientific Chemicals Product Code.: 11499566

Pyridine hydrobromide, 98%, Thermo Scientific Chemicals

Product Code. 11499566
1000 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11499566

Brand: Thermo Scientific Alfa Aesar A19314.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18820-82-1
Molecular Formula C5H6BrN
Molecular Weight (g/mol) 160.014
MDL Number MFCD00012801
InChI Key BBFCIBZLAVOLCF-UHFFFAOYSA-N
Synonym pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at
PubChem CID 87810
IUPAC Name pyridine;hydrobromide
SMILES C1=CC=NC=C1.Br

Specifications

Melting Point 216°C to 220°C
Quantity 1000 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 160.01
Percent Purity 98%
Chemical Name or Material Pyridine hydrobromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252174 Certificate TypeCertificate of Analysis Date10/09/2024 Catalog NumberA19314.30,A19314.18,A19314.0B
Lot Number10245707 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA19314.30,A19314.18,A19314.0B
Lot Number10219827 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19314.30,A19314.18,A19314.0B
Lot Number10194591 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19314.30,A19314.18,A19314.0B
Lot NumberH3562A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19314.30,A19314.18,A19314.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyridine hydrobromide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.