Learn More
Tocris Bioscience™ Pyridostatin pentahydrochloride
Stabilizes G-quadruplexes; induces DNA damage and cell cycle arrest
308.00 €
Specifications
CAS | 1781882-65-2 |
---|---|
Product Type | Pyridostatin pentahydrochloride |
Inhibitors | Pyridostatin pentahydrochloride |
Description
Binds and stabilizes G-quadruplexes, inducing DNA damage and cell cycle arrest (Kd = 490 nM); targets the proto-oncogene Src, reducing Src protein abundance and Src-dependent motility in human breast cancer cells. Also targets telomeric G-quadruplexes, inducing telomerase dysfunction. Activates the DNA-dependent protein kinase catalytic sunbunit (DNA-PKcs).Specifications
1781882-65-2 | |
Pyridostatin pentahydrochloride | |
Pyridostatin pentahydrochloride |