Learn More
Pyridostigmine-d6 bromide, MedChemExpress
MedChemExpress Pyridostigmine D6 bromide is the deuterium labeled Pyridostigmine, which is a parasympathomimetic and a reversible cholinesterase inhibitor.
Brand:  Medchem Express HY-B0207AS
Encompass_Preferred
 View more versions of this product
Product Code. 16400828
  • 452.25 € / 5mg
Explore more special offers
Description
Description
Pyridostigmine D6 bromide is the deuterium labeled Pyridostigmine, which is a parasympathomimetic and a reversible cholinesterase inhibitor.
Specifications
Specifications
Pyridostigmine-d6 bromide
5 mg
Research
DMSO : 50 mg/mL (187.16 mM; Need ultrasonic)
267.15
98.0%
Solid
2375858-08-3
White
C9H7D6BrN2O2
O=C(N(C([2H])([2H])[2H])C([2H])([2H])[2H])OC1=CC=C[N+](C)=C1.[Br-]
267.15
Research