Learn More
Pyridostigmine-d6 bromide, MedChemExpress
MedChemExpress Pyridostigmine D6 bromide is the deuterium labeled Pyridostigmine, which is a parasympathomimetic and a reversible cholinesterase inhibitor.
126.90 € - 716.85 €
Specifications
Chemical Name or Material | Pyridostigmine-d6 bromide |
---|---|
CAS | 2375858-08-3 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C9H7D6BrN2O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16478418
|
Medchem Express
HY-B0207AS |
10 mg |
716.85 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16432089
|
Medchem Express
HY-B0207AS |
1 mg |
126.90 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16400828
|
Medchem Express
HY-B0207AS |
5 mg |
430.65 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pyridostigmine D6 bromide is the deuterium labeled Pyridostigmine, which is a parasympathomimetic and a reversible cholinesterase inhibitor.Specifications
Pyridostigmine-d6 bromide | |
White | |
C9H7D6BrN2O2 | |
O=C(N(C([2H])([2H])[2H])C([2H])([2H])[2H])OC1=CC=C[N+](C)=C1.[Br-] | |
267.15 | |
Research |
2375858-08-3 | |
Research | |
DMSO : 50 mg/mL (187.16 mM; Need ultrasonic) | |
267.15 | |
98.0% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.