missing translation for 'onlineSavingsMsg'
Learn More

Pyrophosphoryl chloride, Thermo Scientific Chemicals

18.00 € - 281.00 €

Chemical Identifiers

CAS 13498-14-1
Molecular Formula Cl4O3P2
Molecular Weight (g/mol) 251.745
MDL Number MFCD00061542
InChI Key CNTIXUGILVWVHR-UHFFFAOYSA-N
Synonym diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, phosphorodichloridic anhydride
PubChem CID 4432411
SMILES O=P(OP(=O)(Cl)Cl)(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358027
View Documents
Thermo Scientific Alfa Aesar
022362.02
0.5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11348027
View Documents
Thermo Scientific Alfa Aesar
022362.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11368027
View Documents
Thermo Scientific Alfa Aesar
022362.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Pyrophosphoryl chloride serves as a reductive chlorination reagent used in the synthesis of quetiapine. It acts as a phosphorylating agent involved in nucleoside synthesis. It plays an important role in Vilsmeier formylation and glyoxylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13498-14-1
251.745
CNTIXUGILVWVHR-UHFFFAOYSA-N
4432411
Cl4O3P2
MFCD00061542
diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, phosphorodichloridic anhydride
O=P(OP(=O)(Cl)Cl)(Cl)Cl

Specifications

101°C (10mmHg)
13498-14-1
Liquid
MFCD00061542
diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, phosphorodichloridic anhydride
CNTIXUGILVWVHR-UHFFFAOYSA-N
251.745
251.76
Air & Moisture Sensitive
Pyrophosphoryl chloride
-17°C
Colorless through Yellow
Cl4O3P2
UN3264
Reacts violently with waterMiscible with most organic solvents.
O=P(OP(=O)(Cl)Cl)(Cl)Cl
4432411
Acrid
1.82g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: I

missing translation for 'einecsNumber' : 236-824-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only