missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ pZerve Cryopreservation Solution
A cryopreservation solution that does not contain dimethyl sulfoxide (DMSO) and serum.
Brand: MP Biomedicals™ 092030346
This item is not returnable.
View return policy
Description
pZerve has been successfully used with a variety of cell types including mast cells, dendritic cells, and mesenchymal cells. It can be used for cells cultured in serum-free medium or with serum-containing medium.

Specifications
Solution | |
Cryopreservation | |
Store at 2° to 8°C |
Cryopreservation Media | |
20 mL | |
pZerve Cryopreservation Media |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ pZerve Cryopreservation Solution > pZerve Cryopreservation Solution, 20mL
Spot an opportunity for improvement?Share a Content Correction