missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Q-PHOENIX-90 Yellow Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas with fire resistance up to 90 minutes.
POA
Specifications
Dimensions (D x W x H) | 615 x 1193 x 1953 mm |
---|---|
Dimensions (D x W x H) Interior | 522 x 1050 x 1647 mm |
Product Type | Safety storage cabinet |
Door Style | Folding |
No. of Doors | 2 |
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
15987406
|
asecos™
3214904033640 |
3 x tray shelf, 1 x perforated insert (365 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15890238
|
asecos™
3214904033635 |
Shelves: 75 kg each, Insert: 75 kg, Sump: 33 L, Maximum load: 600 kg |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937416
|
asecos™
3214904033647 |
4 x drawer (sheet steel powder-coated) |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957416
|
asecos™
3214904033649 |
6 x Drawers 60 kg/29 L each |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Extremely robust design with scratch-proof surface
- Space-saving folding doors, ideal for use in case of narrow passage widths in front of the cabinet
- Continuous anti-trapping door strips on the folding doors offer maximum protection during operation
- Integrated transport base for simple in-house transport, can be moved upright through doors
- Adjustable feet installed in the base allow the safe and fast compensation of minor surface unevenness
- Modern bicolor design and language-free labelling
- Fire resistant up to 90 minutes
- Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
- Locking status indicator (red/green) signals the locking status
- In accordance with EN 14470-1 and TRGS 510
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
- Available interior equipment includes drawers, height-adjustable shelves and bottom collecting sumps
- Triple-hinged doors, safety elements assembled outside the storage compartment for increased protection against corrosion
- Doors stop in any position and self-close in the event of a fire
Specifications
615 x 1193 x 1953 mm | |
Safety storage cabinet | |
2 |
522 x 1050 x 1647 mm | |
Folding |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
asecos™ Q-PHOENIX-90 Yellow Safety Storage Cabinets