Learn More
asecos™ Q-PHOENIX-90 Yellow Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas with fire resistance up to 90 minutes.
Brand:  asecos™ 3214904033647
 View more versions of this product
Product Code. 15937416
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Space saving folding doors: less required space in front of the cabinet while doors are open
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy transport: integrated transport base with removable, optional base cover
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
asecos™ Q-PHOENIX-90 Yellow Safety Storage Cabinets > 4 x drawer (sheet steel powder-coated)