Learn More
Hellma™ Suprasil™ Quartz Micro Absorption Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Suprasil™ Quartz Micro Absorption Cell Cuvettes come with either a lid or PTFE cap to limit evaporation.
Brand:  Hellma™ 100-100-40
 View more versions of this product
Product Code. 11711764
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Very chemically resistant
  • Maintains shape, even at high temperatures up to approximately 1000°C
  • 80% transmission from 320 to 2500nm
Specifications
Specifications
Macro Absorption Cell
Quartz Suprasil
100 mm
45 mm
35,000 μL
9.5mm
200 to 2500 nm
12.5 mm