Learn More
Macherey-Nagel™ Quantofix™ Test Strips
Perform fast, easy analyses on the spot. Macherey-Nagel™ Quantofix™ Test Strips are ready to use semi-quantitative tests that change color depending on the concentration of the analyte in the sample. Read results using the color scale or with the Quantofix™ Relax.
Brand:  Macherey-Nagel™ 91341
 View more versions of this product
Product Code. 11807073
  • 43.56 € / Pack of 100
Edge
Explore more special offers
Description
Description
Perform fast, easy analyses on the spot. Macherey-Nagel™ Quantofix™ Test Strips are ready to use semi-quantitative tests that change color depending on the concentration of the analyte in the sample. Read results using the color scale or with the Quantofix™ Relax.
Quantofix™ test strips meet all requirements of a modern rapid test. Analytical professionals as well as occasional testers appreciate the test strips for the fast and easy analysis directly at the point of interest. Often, these tests are used to quickly check whether important parameters are in the desired range. They deliver an immediate result and thus enable a fast response.
  • Rapid
    • Just Dip & Read
    • Results within seconds
    • Always ready for use
  • Easy
    • No calibration
    • No maintenance
    • No accessories
  • Reliable
    • Desiccant in the stopper for optimal protection of the strips against humidity
    • Color chart confirmed with traceable standards
    • Automatic evaluation with Quantofix™ Relax for safe documentation
Specifications
Specifications
CE for disinfection/medical products
Peracetic Acid 500
Semi Quantitative Test Strips
Determination of Peracetic Acid
0, 50, 100, 200, 300, 400, 500mg/L Peracetic Acid
Yellow to Green
Peracetic Acid
Redox Reaction
50 - 500mg/L Peracetic Acid
30 sec.
Product Suggestions
Product Suggestions
Videos
SDS Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Quantofix™ Test Strips > Graduations: 0 to 500mg/L