Learn More
Macherey-Nagel™ Quantofix™ Test Strips
Perform fast, easy analyses on the spot. Macherey-Nagel™ Quantofix™ Test Strips are ready to use semi-quantitative tests that change color depending on the concentration of the analyte in the sample. Read results using the color scale or with the Quantofix™ Relax.
28.85 € - 43.00 €
Products 4
Product Code Brand Description Range Color Code For Use With (Application) Price Quantity & Availability  
Product Code Brand Description Range Color Code For Use With (Application) Price Quantity & Availability  
15171136
View Documents
Macherey-Nagel™
91350
Silver 0, 1, 2, 3, 5, 7, 10g/L Ag Yellow to Brown Determination of silver
29.25 €
Each
11807073
View Documents
Macherey-Nagel™
91341
Peracetic Acid 500 0, 50, 100, 200, 300, 400, 500mg/L Peracetic Acid Yellow to Green Determination of Peracetic Acid
43.00 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10078222
View Documents
Macherey-Nagel™
91336
LubriCheck 0, 15, 50, 75, 130, 200mmol/L KOH Yellow to Blue Determination of Potassium Hydroxide
38.80 €
Pack of 100
In Stock
to see stock.
Add to basket
12691515
View Documents
Macherey-Nagel™
91330
Total Iron 1000 0, 5, 20, 50, 100, 250, 500, 1000mg/L Fe2+/3+ White to Dark Red Determination of Total Iron 1000
28.85 €
Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
Perform fast, easy analyses on the spot. Macherey-Nagel™ Quantofix™ Test Strips are ready to use semi-quantitative tests that change color depending on the concentration of the analyte in the sample. Read results using the color scale or with the Quantofix™ Relax.
Quantofix™ test strips meet all requirements of a modern rapid test. Analytical professionals as well as occasional testers appreciate the test strips for the fast and easy analysis directly at the point of interest. Often, these tests are used to quickly check whether important parameters are in the desired range. They deliver an immediate result and thus enable a fast response.
  • Rapid
    • Just Dip & Read
    • Results within seconds
    • Always ready for use
  • Easy
    • No calibration
    • No maintenance
    • No accessories
  • Reliable
    • Desiccant in the stopper for optimal protection of the strips against humidity
    • Color chart confirmed with traceable standards
    • Automatic evaluation with Quantofix™ Relax for safe documentation
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 2.5 Years