Learn More
Hellma™ Quartz Suprasil Ultra-Micro Absorption Cells
Brand:  Hellma™ 105-201-15-40
 View more versions of this product
Product Code. 10648972
  • 559.00 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Ultra-micro Absorption Cell
Quartz Suprasil
10 mm
Photometers and Spectrophotometers
0.7 to 5 μL
1 Each
45 mm
12.5 mm