Learn More
Hellma™ Quartz Suprasil Ultra-Micro Absorption Cells
244.00 € - 769.00 €
Specifications
Type | Ultra-micro Absorption Cell |
---|---|
Material | Quartz Suprasil |
Width (Metric) | 12.5 mm |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10648972
|
Hellma™
105-201-15-40 |
0.7 to 5 μL |
559.00 €
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
10446491
|
Hellma™
105-251-15-40 |
100 μL |
769.00 €
Each |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
11756214
|
Hellma™
105-200-15-40 |
300 μL | N/A | ||||||
Specifications
Ultra-micro Absorption Cell | |
Quartz Suprasil | |
12.5 mm |