Learn More
Hellma™ Quartz Suprasils Macro Absorption Cells
Brand:  Hellma™ 100-2-40
 View more versions of this product
Product Code. 15198444
  • 187.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Macro Absorption Cell
Quartz Suprasil
1 Each
45 mm
26 μL
9.5mm
2 mm
12.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products