Learn More
Hellma™ Quartz UV Flow Cell
Brand:  Hellma™ 170700-0.5-40
 View more versions of this product
Product Code. 12663006
  • 1205.00 € / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
Specifications
UV Flow Cell
31 μL
Quartz
0.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hellma™ Quartz UV Flow Cell > Capacity: 31μL