Learn More
Hellma™ Quartz UV Flow Cell
826.00 € - 1205.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
12791705
|
Hellma™
170700-0.2-40 |
12.4 μL |
1205.00 €
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
||||||
12637201
|
Hellma™
170-000-1-40 |
120 μL |
826.00 €
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
||||||
12663006
|
Hellma™
170700-0.5-40 |
31 μL |
1205.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
Specifications
UV Flow Cell | |
Quartz |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hellma™ Quartz UV Flow Cell