missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Quartz Wool

Filter aid

31.20 € - 215.07 €

Specifications

For Use With (Application) Filter aid
Material Quartz

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15575086
SDS View Documents
Thermo Scientific Chemicals
393610100
10 g N/A N/A N/A
15585086
SDS View Documents
Thermo Scientific Chemicals
393611000
100 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Filter aid
Quartz
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Quartz wool, coarse

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Quartz Wool

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.