missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit and Quant-it Assay Kit Standards

Achieve precise quantitation of dsDNA or RNA samples with stand-alone Qubit nucleic acid standards.

Brand:  Invitrogen™ Q33233

57.57 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 15880210

  • 79.25 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Achieve precise quantitation of dsDNA or RNA samples with stand-alone Qubit nucleic acid standards. The Qubit standards can be used in conjunction with different Qubit or Quant-iT nucleic acid quantitation assay kits and offer stand-alone versions of the kit standard for performing additional assays.

Qubit 1X dsDNA HS Assay Lambda Standard

Qubit 1X dsDNA HS Assay Lambda Standard can be used with either Qubit 1X dsDNA HS Assay Kits, Qubit dsDNA HS Assay Kits, or their corresponding Quant-iT Assay Kits. The standards are supplied at the same concentration as the highest concentration dsDNA standard (10 ng/μL) in the Quant-iT Assay Kits and can be diluted into the standard series.

Qubit 1X dsDNA BR Standards

Qubit 1X dsDNA BR (Broad Range) Standards can be used with either Qubit 1X dsDNA BR Assay Kits, Qubit dsDNA BR Assay Kits, or their corresponding Quant-iT Assay Kits. The Qubit 1X dsDNA BR Standards includes standards at two concentrations, 0 and 100 ng/μL. The high standard corresponds to the same concentration as the highest concentration dsDNA standard in the Quant-iT Assay Kits and can be diluted into the standard series.

Qubit RNA IQ Assay Standards

Qubit RNA IQ Assay RNA standards are designed for use with the Qubit RNA IQ Assay Kit. Sufficient amounts of the small and large and/or structured RNA standard are supplied for generating an IQ standard curve for up to 500 samples. The RNA IQ standards are designed to measure RNA degradation and are not interchangeable with the standards from other RNA quantitation assays, including the Qubit RNA HS, BR, or XR assay kits.

Note: Use thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex fluorometer.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Qubit™ dsDNA HS Assay – Lambda DNA Standard, 5 mL at 10 ng/μL in TE buffer
Store at 2°C to 8°C
Qubit and Quant‐iT dsDNA HS Assays and 1 x dsDNA Assays
Fewer resources used, Less waste
1X dsDNA HS Assay Lambda Standard
Room Temperature
10 ng/μL
Qubit Fluorometer or fluorescence microplate reader
Qubit
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2980688 Certificate TypeCertificate of Analysis Date03/09/2024 Catalog NumberQ33233
Lot Number2896404 Certificate TypeCertificate of Analysis Date30/05/2024 Catalog NumberQ33233
Lot Number2896403 Certificate TypeCertificate of Analysis Date15/02/2024 Catalog NumberQ33233
Lot Number2881057 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog NumberQ33233
Lot Number2862023 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog NumberQ33233
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Qubit and Quant-it Assay Kit Standards > Type: 1X dsDNA HS Assay Lambda Standard

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.