missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Double Surface Davies Condenser

Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Double Surface Davies Condenser is highly efficient with a large cooling surface. The inner cooling surface prevents vibration with use of the baffle effect, while the outer cooling surface prevents creep with low boiling point liquids. Includes detachable plastic screw thread connectors for easy and safe tubing attachment.

Brand:  Quickfit C5/12/SC

109.80 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11301865

  • 127.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Double Surface Davies Condenser is highly efficient with a large cooling surface. The inner cooling surface prevents vibration with use of the baffle effect, while the outer cooling surface prevents creep with low boiling point liquids. Includes detachable plastic screw thread connectors for easy and safe tubing attachment.

  • Highly efficient condenser with large cooling surface
  • Inner cooling surface gives 'baffle' effect
  • Outer cooling surface prevents 'creep' with low boiling point liquids
  • With detachable plastic screw thread connectors for easy and safe attachment of flexible tubing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Surface area x 102m2: 1.7
295 mm
Borosilicate Glass
160 mm
19/26
Davies Condenser
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers