Learn More
Quickfit™ Borosilicate Glass Double Surface Davies Condenser
Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Double Surface Davies Condenser is highly efficient with a large cooling surface. The inner cooling surface prevents vibration with use of the baffle effect, while the outer cooling surface prevents creep with low boiling point liquids. Includes detachable plastic screw thread connectors for easy and safe tubing attachment.
131.00 € - 297.00 €
Product Code | Brand | Length (Metric) | Length (Metric) Jacket | Standard Taper Joint No | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Length (Metric) Jacket | Standard Taper Joint No | Description | Price | Quantity & Availability | ||||||
11301865
|
Quickfit
C5/12/SC |
295 mm | 160 mm | 19/26 | Surface area x 102m2: 1.7 |
134.00 €
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
11391855
|
Quickfit
C5/11/SC |
275 mm | 160 mm | 14/23 | Surface area x 102m2: 1.7 |
131.00 €
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
11311865
|
Quickfit
C5/22/SC |
340 mm | 205 mm | 19/26 | Surface area x 102m2: 2.3 |
131.00 €
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
11321865
|
Quickfit
C5/23/SC |
345 mm | 205 mm | 24/29 | Surface area x 102m2: 3.0 |
181.00 €
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
10074754
|
Quickfit
C5/24/SC |
365 mm | 205 mm | 29/32 | Surface area x 102m2: 3.0 |
139.00 €
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
12408806
|
Quickfit
C5/13/SC |
300 mm | 160 mm | 24/29 | Surface area x 102m2: 2.3 |
131.00 €
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
15567780
|
Quickfit
CX5/23/SC |
350 mm | 205 mm | 24/29 | Surface area x 102m2: 3.0 | N/A | N/A | N/A | |||||
15577780
|
Quickfit
CX5/25/SC |
360 mm | 205 mm | 34/35 | Surface area x 102m2: 3.0 |
297.00 €
Pack of 2 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
Description
Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Double Surface Davies Condenser is highly efficient with a large cooling surface. The inner cooling surface prevents vibration with use of the baffle effect, while the outer cooling surface prevents creep with low boiling point liquids. Includes detachable plastic screw thread connectors for easy and safe tubing attachment.
- Highly efficient condenser with large cooling surface
- Inner cooling surface gives 'baffle' effect
- Outer cooling surface prevents 'creep' with low boiling point liquids
- With detachable plastic screw thread connectors for easy and safe attachment of flexible tubing
Specifications
Borosilicate Glass |