Learn More
Quickfit™ Borosilicate Glass Liebig Condenser
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
Brand:  Quickfit C1/00
83.82 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 11391845
  • 112.00 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
  • Complies with DIN 12576
  • Popular water-jacketed condenser suitable for distillation and reflux applications
  • With detachable plastic screwthread connectors for easy and safe attachment of rubber hose
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN 12576
135 mm
Borosilicate Glass
Suitable for distillation and reflux applications
10/19
Liebig Condenser
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Liebig Condenser > Cone Size: 10/19; Length: 135mm; Effective Length: 65mm