Learn More
Quickfit™ Borosilicate Glass Liebig Condenser
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
Brand:  Quickfit C1/22/SC
48.19 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 12049652
  • 66.05 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
  • Complies with DIN 12576
  • Popular water-jacketed condenser suitable for distillation and reflux applications
  • With detachable plastic screwthread connectors for easy and safe attachment of rubber hose
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN 12576
Suitable for distillation and reflux applications
530 mm
Borosilicate Glass
Surface Area X 102m2: 2.00
400 mm
19/26
Liebig Condenser
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Liebig Condenser > Cone Size: 19/26; Length: 530mm; Effective Length: 398mm