Learn More
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
89.50 € - 882.00 €
Specifications
Certifications/Compliance | ISO 1773, ISO 4797 |
---|---|
Material | Borosilicate Glass |
Neck Style | Short |
Bottom Shape | Flat |
Product Code | Brand | Capacity (Metric) | Height (Metric) | Socket Joint | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Height (Metric) | Socket Joint | Price | Quantity & Availability | ||||||
12498936
|
Quickfit
FF1L/3US |
1000 mL | 170 mm | 24/29 |
89.50 €
Pack of 2 |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
12498926
|
Quickfit
FF100/2S |
100 mL | 95 mm | 19/26 |
133.00 €
Pack of 5 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
12418936
|
Quickfit
FF100/3S |
100 mL | 103 mm | 24/29 |
170.00 €
Pack of 5 |
Estimated Shipment: 26-06-2025
Log in to see stock. |
||||||
12428936
|
Quickfit
FF100/4S |
100 mL | 103 mm | 29/32 |
133.00 €
Pack of 5 |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
10243930
|
Quickfit
FF150/3S |
150 mL | 110 mm | 24/29 |
151.00 €
Pack of 5 |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
10697445
|
Quickfit
FF150/5S |
150 mL | 115 mm | 34/35 |
155.00 €
Pack of 5 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
12428946
|
Quickfit
FF250/2S |
250 mL | 130 mm | 19/26 |
225.00 €
Pack of 5 |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
10309754
|
Quickfit
FF250/4S |
250 mL | 130 mm | 29/32 |
128.00 €
Pack of 5 |
In Stock
Log in to see stock. ![]() |
||||||
10831041
|
Quickfit
FF250/5S |
250 mL | 130 mm | 34/35 |
219.00 €
Pack of 5 |
In Stock
Log in to see stock. ![]() |
||||||
12509885
|
Quickfit
FF500/3S |
500 mL | 140 mm | 24/29 |
167.00 €
Pack of 5 |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
12418956
|
Quickfit
FF500/4S |
500 mL | 145 mm | 29/32 |
178.00 €
Pack of 5 |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
10851201
|
Quickfit
FF50/2S |
50 mL | 85 mm | 19/26 |
125.00 €
Pack of 5 |
Estimated Shipment: 26-06-2025
Log in to see stock. |
||||||
12661992
|
Quickfit
FF50/3S |
50 mL | 85 mm | 24/29 |
121.00 €
Pack of 5 |
Estimated Shipment: 26-06-2025
Log in to see stock. |
||||||
11980885
|
Quickfit
FF250/3S |
250 mL | 130 mm | 24/29 |
142.00 €
Pack of 5 |
In Stock
Log in to see stock. ![]() |
||||||
15527840
|
Quickfit
FF2L/5S |
2000 mL | 215 mm | 34/35 |
434.00 €
Pack of 5 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
12408946
|
Quickfit
FF1L/4S |
1000 mL | 187 mm | 29/32 |
89.50 €
Pack of 2 |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
15567830
|
Quickfit
FF150/2S |
150 mL | 105 mm | 19/26 |
605.00 €
Pack of 20 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
15577830
|
Quickfit
FF150/4S |
150 mL | 110 mm | 29/32 |
605.00 €
Pack of 20 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
15517840
|
Quickfit
FF2L/4S |
2000 mL | 230 mm | 29/32 |
465.00 €
Pack of 5 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
15547840
|
Quickfit
FF500/2S |
500 mL | 135 mm | 19/26 |
662.00 €
Pack of 20 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
15567840
|
Quickfit
FF500/5S |
500 mL | 155 mm | 34/35 |
882.00 €
Pack of 20 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
15557830
|
Quickfit
FF100/5S |
100 mL | 103 mm | N/A | N/A | N/A | ||||||
15597830
|
Quickfit
FF1L/5S |
1000 mL | 175 mm | 34/35 |
528.00 €
Pack of 10 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
15507840
|
Quickfit
FF2L/3S |
2000 mL | 205 mm | 24/29 |
470.00 €
Pack of 5 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
15537840
|
Quickfit
FF4L/7S |
4 L | 260 mm | N/A | N/A | N/A | ||||||
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
- Complies with ISO 1773 and ISO 4797
- With highest quality Quickfit™ ground glass socket
- Borosilicate glass for highest resistance to thermal shock and chemical corrosion
- Flat base allows use with hotplates
Specifications
ISO 1773, ISO 4797 | |
Borosilicate Glass | |
Short | |
Flat |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask