Learn More
Quickfit™ Borosilicate Glass Vigreux Fractionating Column
Popular and easy-to-use fractionating column using a combination of horizontal and angled indentations. Quickfit™ Borosilicate Glass Vigreux Fractionating Column has a significantly increased column surface area for greater efficiency.
Brand: Quickfit™ FC3/13
132.05 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
Popular and easy-to-use fractionating column using a combination of horizontal and angled indentations. Quickfit™ Borosilicate Glass Vigreux Fractionating Column has a significantly increased column surface area for greater efficiency.
- Popular, easy to use column
- Vigreux type with combination of horizontal and angled indentations
- Horizontal indentations ensure intimate vapour and liquid contact
- Angled indentations redistribute liquid from walls to centre of column

Specifications
360mm | |
Vigreux Fractionating Column | |
Borosilicate Glass | |
ISO 3585 |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.