missing translation for 'onlineSavingsMsg'
Learn More

Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask

Manufactured from borosilicate glass. Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask's wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues.

Brand:  Quickfit FR10LF

 View more versions of this product

Product Code. 11971279

  • 388.00 € / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from borosilicate glass

  • With large diameter Pyrex Labware™ Quickfit™ flat-flange joint
  • For use with Pyrex Labware™ Quickfit™ flat-flange lids and retaining clips
  • Wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 L
For use with Quickfit™ flat-flange lids and retaining clips
Wide
Borosilicate Glass
320 mm
Round
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask > Capacity: 10000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.