Learn More
Quickfit™ Glass Büchner Funnel with Glass Sintered Disc
Used in place of conventional filter papers that would be destroyed by acids and alkalis. Quickfit™ Glass Büchner Funnel with Sintered Glass Disc is manufactured from chemical-resistant borosilicate glass. Integral ground glass cone on stem connects to Quickfit™ Büchner flasks and vessels.
Brand:  Quickfit SF3A32
 View more versions of this product
Product Code. 12425386
  • 158.00 € / Each
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Used in place of conventional filter papers that would be destroyed by acids and alkalis. Quickfit™ Glass Büchner Funnel with Sintered Glass Disc is manufactured from chemical-resistant borosilicate glass. Integral ground glass cone on stem connects to Quickfit™ Büchner flasks and vessels.
  • Büchner funnel with glass sintered disc
  • Borosilicate glass construction for high resistance to chemical attack
  • Useful where conventional filter papers would be destroyed by acids and alkalis
  • Integral ground glass cone on stem for connecting to Quickfit™ Büchner flasks etc
TRUSTED_SUSTAINABILITY
Specifications
Specifications
125 mL
19/26
55mm
3
Buchner Funnel
Borosilicate Glass
Bases or acids that can destroy conventional filter papers
Sintered glass disc, ground glass cone
16 to 40μm