missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Glass Conical Funnel with Glass Sintered Disc

Constructed of chemically resistant borosilicate glass. Quickfit™ Glass Conical Funnel with Glass Sintered Disc is resistant to most corrosive reagents such as ammonia and sulphuric acid. Conical shape with glass sintered disc eliminates the need for filter papers. Integral ground glass cone on the stem connects to Quickfit™ Büchner flasks and vessels.

Brand:  Quickfit SF4A32

91.59 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.


 View more versions of this product

Product Code. 12455386

  • 109.00 € / Each
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Constructed of chemically resistant borosilicate glass. Quickfit™ Glass Conical Funnel with Glass Sintered Disc is resistant to most corrosive reagents such as ammonia and sulphuric acid. Conical shape with glass sintered disc eliminates the need for filter papers. Integral ground glass cone on the stem connects to Quickfit™ Büchner flasks and vessels.

  • Conical shape with glass sintered disc
  • Resistant to most corrosive reagents - Ammonia, sulfuric acid, etc
  • With integral ground glass cone on the stem for connecting to Quickfit™ Büchner flasks
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Borosilicate Glass
For connecting to Quickfit™ Buchner flasks
Sintered glass disc
16 to 40μm
19/26
20mm
3
Filter Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Quickfit™ Glass Conical Funnel with Glass Sintered Disc > Disk Diameter: 20mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.