Learn More
Quickfit™ Glass Conical Funnel with Glass Sintered Disc
Constructed of chemically resistant borosilicate glass. Quickfit™ Glass Conical Funnel with Glass Sintered Disc is resistant to most corrosive reagents such as ammonia and sulphuric acid. Conical shape with glass sintered disc eliminates the need for filter papers. Integral ground glass cone on the stem connects to Quickfit™ Büchner flasks and vessels.
104.00 € - 109.00 €
Specifications
Material | Borosilicate Glass |
---|---|
Cone Size | 19/26 |
For Use With (Application) | For connecting to Quickfit™ Buchner flasks |
Includes | Sintered glass disc |
Porosity | 3 |
Product Code | Brand | Diameter (Metric) Disc | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Disc | Price | Quantity & Availability | ||||||
12455386
|
Quickfit
SF4A32 |
20mm |
109.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
12018483
|
Quickfit
SF4B32 |
30mm |
104.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
Description
Constructed of chemically resistant borosilicate glass. Quickfit™ Glass Conical Funnel with Glass Sintered Disc is resistant to most corrosive reagents such as ammonia and sulphuric acid. Conical shape with glass sintered disc eliminates the need for filter papers. Integral ground glass cone on the stem connects to Quickfit™ Büchner flasks and vessels.
- Conical shape with glass sintered disc
- Resistant to most corrosive reagents such as-ammonia and sulfuric acid
- With integral ground glass cone on the stem for connecting to Quickfit™ Büchner flasks
Specifications
Borosilicate Glass | |
For connecting to Quickfit™ Buchner flasks | |
3 | |
Filter Funnel |
19/26 | |
Sintered glass disc | |
16 to 40μm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.