Learn More
Quickfit™ PTFE Screwthread Washers
Secure screwcaps to glass screwthreads. QuickFit™ screwthread washers for QuickFit™ screwthreads ensure that the cap fits correctly. Washers come in a variety of sizes.
23.00 € - 42.95 €
Specifications
Material | PTFE |
---|---|
For Use With | For Quickfit™ Glass screw threads |
Product Type | Washer |
Product Code | Brand | Thread Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | ||||||
12939283
|
Quickfit
QW13/6 |
13 |
25.60 €
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
12959283
|
Quickfit
QW24/6 |
24 |
42.95 €
Pack of 10 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
12969283
|
Quickfit
QW24/9 |
24 | N/A | N/A | N/A | |||||
12999283
|
Quickfit
QW28/11 |
28 |
36.70 €
Pack of 10 |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
10538220
|
Quickfit
QW18/7 |
18 |
23.00 €
Pack of 10 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
11332085
|
Quickfit
QW24/11 |
24 |
41.30 €
Pack of 10 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
12989283
|
Quickfit
QW28/6 |
28 | N/A | N/A | N/A | |||||
Description
Secure screwcaps to glass screwthreads. QuickFit™ screwthread washers for QuickFit™ screwthreads ensure that the cap fits correctly. Washers come in a variety of sizes.
- For use with Quickfit™ glass screwthreads
- Melamine screwcaps with aperture at the top
- Silicone rubber rings with PTFE washers
- To determine what components are required: i) Check the size of the glass screw thread (size designation refers to O. D. of thread inmm) ii) Check the O. D. of the sampling tube, thermometer, probe etc. , that is to be passed through the screw cap.
Specifications
PTFE | |
For Quickfit™ Glass screw threads | |
Washer |