Learn More
Quickfit™ Reduction Socket Adapter
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
41.05 € - 227.00 €
Specifications
For Use With (Equipment) Dissimilar sized ground Glass joint
Includes Small socket, large cone
Material Borosilicate Glass
Product Type Reduction adapter
View More Specs
Products 18
Product Code Brand Cone Size Standard Taper Joint No. Product Type Price Quantity & Availability  
Product Code Brand Cone Size Standard Taper Joint No. Product Type Price Quantity & Availability  
10203720
View Documents
Quickfit
DA12
Cone Size: 19/26 14/23 Reduction adapter
62.45 €
Pack of 2
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
10520681
View Documents
Quickfit
DA13
Cone Size: 24/29 14/23 Reduction adapter
60.95 €
Pack of 2
In Stock
to see stock.
Edge
Add to basket
12634891
View Documents
Quickfit
DA23
Cone Size: 24/29 19/26 Reduction adapter
60.40 €
Pack of 2
In Stock
to see stock.
Edge
Add to basket
12418876
View Documents
Quickfit
DA14
Cone Size: 29/32 14/23 Reduction adapter
58.19 €
Pack of 2
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
12520337
View Documents
Quickfit
DA01
Cone Size: 14/23 10/19 Reduction adapter
42.00 €
Pack of 2
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10765861
View Documents
Quickfit
DA24
Cone Size: 29/32 19/26 Reduction adapter
60.93 €
Pack of 2
In Stock
to see stock.
Add to basket
12448876
View Documents
Quickfit
DA25
Cone Size: 34/35 19/26 Reduction adapter
53.05 €
Pack of 2
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
12989253
View Documents
Quickfit
DA35
Cone Size: 34/35 24/29 Reduction adapter
52.25 €
Pack of 2
In Stock
to see stock.
Edge
Add to basket
12654891
View Documents
Quickfit
DA34
Cone Size: 29/32 24/29 Reduction adapter
59.05 €
Pack of 2
In Stock
to see stock.
Edge
Add to basket
12478876
View Documents
Quickfit
DA45
Cone Size: 34/35 29/32 Reduction adapter
120.64 €
Pack of 2
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
12050103
View Documents
Quickfit
DA58
Cone Size: 50/42 34/35 Reduction adapter
41.05 €
Each
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
15212806
View Documents
Quickfit
DA57
Cone Size: 45/40 34/35 Reduction adapter
207.00 €
Each
In Stock
to see stock.
Edge
Add to basket
15597790
View Documents
Quickfit
DA36
Cone Size: 40/38 24/29 Reduction adapter
177.00 €
Pack of 5
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
15507800
View Documents
Quickfit
DA37
Cone Size: 45/40 24/29 Reduction adapter
207.00 €
Pack of 5
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
15517800
View Documents
Quickfit
DA38
Cone Size: 50/42 24/29 Reduction adapter
216.00 €
Pack of 5
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
15527800
View Documents
Quickfit
DA39
Cone Size: 55/44 24/29 Reduction adapter This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15537800
View Documents
Quickfit
DA47
Cone Size: 45/40 29/32 Reduction adapter
227.00 €
Pack of 5
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15547800
View Documents
Quickfit
DA56
Cone Size: 40/38 34/35 Reduction adapter
207.00 €
Pack of 5
Estimated Shipment: 29-08-2025
to see stock.
Add to basket
Description
Description
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
  • Useful for the connection of dissimilar sized ground glass joints
  • Small sized socket at the top, large sized cone at the bottom
Specifications
Specifications
Dissimilar sized ground Glass joint
Small socket, large cone
Borosilicate Glass
Reduction adapter
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Quickfit™ Reduction Socket Adapter