Learn More
Quickfit™ Reduction Socket Adapter
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
Brand: Quickfit DA13
33.45 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
- Useful for the connection of dissimilar sized ground glass joints
- Small sized socket at the top, large sized cone at the bottom
Specifications
Cone Size: 24/29 | |
Small socket, large cone | |
Reduction adapter |
Dissimilar sized ground Glass joint | |
Borosilicate Glass | |
14/23 |