Learn More
IKA™ R 1401 Propeller Stirrer, 3-Bladed
The R 1401 propeller head is designed for the use with the overhead stirrers EUROSTAR 20 high speed digital/control.
Brand: IKA™ 0001242900
Description
It is ideally suited for applications which work with low to medium mixing viscosities that require axial flow pattern at high speed with minimal temperature increase.Order Info
R 1401 needs to be attached to the precision shaft R 6000 in order to operate (must be ordered separately).

Specifications
Working range of 1 to 30L | |
55 mm | |
High speed stirring |
2.2 in. | |
EUROSTAR 20 high speed digital mixers | |
Propeller |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.