missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CD155/PVR His-tag Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 2530-CD-050

273.86 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18128380

  • 358.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its binding ability in a functional ELISA. When Recombinant Human CD155/PVR is coated at 2.5 μg/mL (100 μL/well), the concentration of Recombinant Human TIGIT Fc Chimera (Catalog # 7898-TGB ) that produces 50% optimal binding response is 40-200 ng/mL.
Specifications

Specifications

AAH15542
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
M.W. (Observed): 62-70 kDa, reducing conditions; M.W. (theoretical): 35.3 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
PVR
Recombinant
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Binding Activity
5817
CD155/PVR
Mouse myeloma cell line, NS0-derived human CD155/PVR protein Gly27-Asn343, with a C-terminal 6-His tag
<0.10 EU / 1 μg of the protein by the LAL method.
Unconjugated
Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
CD155/PVR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers