missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CD155/PVR His-tag Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

358.00 €

Specifications

Accession Number AAH15542
For Use With (Application) Binding Activity
Gene ID (Entrez) 5817
Molecular Weight (g/mol) M.W. (Observed): 62-70 kDa, reducing conditions; M.W. (theoretical): 35.3 kDa
Name CD155/PVR
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18128380
View Documents
R&D Systems™
2530-CD-050
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
358.00 €
50µg
Please to purchase this item. Need a web account? Register with us today!
18138020
View Documents
R&D Systems™
2530-CD-050/CF
Supplied as a 0.2 μm filtered solution in PBS.
358.00 €
50µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Measured by its binding ability in a functional ELISA. When Recombinant Human CD155/PVR is coated at 2.5 μg/mL (100 μL/well), the concentration of Recombinant Human TIGIT Fc Chimera (Catalog # 7898-TGB ) that produces 50% optimal binding response is 40-200 ng/mL.
Specifications

Specifications

AAH15542
5817
CD155/PVR
Mouse myeloma cell line, NS0-derived human CD155/PVR protein Gly27-Asn343, with a C-terminal 6-His tag
PVR
Recombinant
CD155/PVR
Binding Activity
M.W. (Observed): 62-70 kDa, reducing conditions; M.W. (theoretical): 35.3 kDa
50 μg
<0.10 EU / 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Videos
SDS
Documents

Documents

Product Certifications