Learn More
Tuttnauer R.PCR Software
PC software for generating reports which include: graph of the cycle data, numeric data of the cycle, copy of cycle print-outs, table of all measured values, table of all parameters and option to save all reports in PDF file format.
Brand:  Tuttnauer ADD222-0461
Product Code. 17879528
  • 615.00 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • PC software for generating reports which include: graph of the cycle data, numeric data of the cycle, copy of cycle print-outs, table of all measured values, table of all parameters and option to save all reports in PDF file format.
The software allows 2 modes of operation:
  • Online mode:
    • Up to 8 autoclaves may be accessed using a PC within the organization via an Ethernet connection.
    • These autoclaves can each be remotely monitored at the same time showing all reports and graphs.
    • History files are automatically downloaded from the autoclave.
  • Offline mode:
    • Data is transferred from the autoclave using a standard USB memory device.
    • This data can then be viewed with the report generator software on a PC.
Specifications
Specifications
Software
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tuttnauer R.PCR Software > R.PCR Software